missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Holder For B-411 and E-416 Tubes

Brand:  BUCHI 043039

Additional Details : Weight : 1.22000kg

Product Code. 10686903

  • € 488.00 / Each
Estimated Shipment: 05-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Test Tube Rack
6
1 Pack
48 mm
Tubes (30 mL), Tubes (48 mm)
B-411 and E-416
1 Pack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers