missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOMER1 Antibody (2F7), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50605B
This item is not returnable.
View return policy
Description
HOMER1 Monoclonal antibody specifically detects HOMER1 in Drosophila samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| HOMER1 | |
| Monoclonal | |
| Biotin | |
| HOMER, homer homolog 1 (Drosophila), homer-1, HOMER1B, HOMER-1B, HOMER1C, SYN47neuronal immediate early gene, 1 | |
| GST-Drosophila HOMER1 (full-length) fusion protein | |
| 0.1 mL | |
| Neuroscience, Neurotransmission | |
| 9456 | |
| Store at 4°C in the dark. |
| Immunofluorescence | |
| 2F7 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Drosophila | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction