missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOMER1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99478AF750
This item is not returnable.
View return policy
Description
HOMER1 Polyclonal antibody specifically detects HOMER1 in Human samples. It is validated for Western Blot
Specifications
| HOMER1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 750 | |
| HOMER, homer homolog 1 (Drosophila), homer-1, HOMER1B, HOMER-1B, HOMER1C, SYN47neuronal immediate early gene, 1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human HOMER1. | |
| 0.1 mL | |
| Neuroscience, Neurotransmission | |
| 9456 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction