missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOMER1 Monoclonal Antibody (OTI3G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811741S
This item is not returnable.
View return policy
Description
HOMER1 Monoclonal antibody specifically detects HOMER1 in Human samples. It is validated for Western BlotSpecifications
| HOMER1 | |
| Monoclonal | |
| 0.5 to 1.0 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UL15 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9529 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI3G5 | |
| Unconjugated | |
| BAG5 | |
| HOMER; homer homolog 1; homer homolog 1 (Drosophila); homer protein homolog 1; homer scaffolding protein 1; homer, neuronal immediate early gene, 1; Homer1; homer-1; HOMER1A; HOMER1B; HOMER1C; HOMER1F; immediate early gene protein Homer1A; PSD-Zip45; scaffold protein; SYN47; VASP/Ena-related gene up-regulated during seizure and LTP 1; Ves-1; Vesl; Vesl1; Vesl-1 | |
| Human recombinant protein fragment corresponding to amino acids 290-440 of human BAG5 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction