missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOMER1 Monoclonal Antibody (OTI3G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811741
This item is not returnable.
View return policy
Description
Homer 1 (Homer protein homolog 1) belongs to the Homer family of proteins that regulate signal transduction through, and the trafficking of, glutamate receptors, as well as maintain and regulate extracellular glutamate levels in corticolimbic brain regions. The Homer 1 is enriched at excitatory synapses and binds group 1 metabotropic glutamate receptors (mGluRs).Specifications
| HOMER1 | |
| Monoclonal | |
| Unconjugated | |
| HOMER1 | |
| HOMER, HOMER1A, HOMER1B, HOMER1C, SYN47, Ves-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 26556, 29546, 9456 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI3G5 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q86YM7, Q9Z214, Q9Z2Y3 | |
| HOMER1 | |
| Human recombit protein fragment corresponding to amino acids 1-354 of human HOMER1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction