missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX17 Monoclonal Antibody (OTI7H5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812124
This item is not returnable.
View return policy
Description
Cytochrome c oxidase 17 (COX17) is a nuclear gene encoding an assembly protein necessary for proper COX apoenzyme-dependent mitochondrial respiration. The mammalian COX apoenzyme is a heteromer consisting of three mitochondrial genes that encode catalytic subunits and several nuclear genes that encode structural subunits. COX17 influences the recruitment of copper to the mitochondria for incorporation into the COX apoenzyme.Specifications
| COX17 | |
| Monoclonal | |
| Unconjugated | |
| COX17 | |
| COX17, cytochrome c oxidase copper chaperone | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10063 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7H5 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14061 | |
| COX17 | |
| Human recombit protein fragment corresponding to amino acids 2-63 of human COX17 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction