Learn More
HOMER2 Monoclonal Antibody (OTI2D5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA806310S
Encompass_Preferred
 View more versions of this product
Product Code. 17586433
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
HOMER2 Monoclonal antibody specifically detects HOMER2 in Human samples. It is validated for Western Blot
Specifications
Specifications
HOMER2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q96E11
Mouse
Affinity chromatography
RUO
92399
-20° C, Avoid Freeze/Thaw Cycles
IgG2a
Western Blot
OTI2D5
Unconjugated
Mrrf
9330120H11Rik; A VESL-2; ACPD; AW539445; CPD; Cupidin; DFNA68; homer 2; homer homolog 2; homer homolog 2 (Drosophila); homer homolog 3; homer protein homolog 2; homer scaffold protein 2; homer scaffolding protein 2; homer, neuronal immediate early gene, 2; HOMER2; homer-2; VASP/Ena-related gene up-regulated during seizure and LTP 2; Vesl2; Vesl-2
Human recombinant protein fragment corresponding to amino acids 56-262 of human MRRF produced in E.coli.
30 μL