missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Homez Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00057594-B01P
This item is not returnable.
View return policy
Description
Homez Polyclonal antibody specifically detects Homez in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Homez | |
| Polyclonal | |
| Western Blot 1:500, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| homeobox and leucine zipper encoding, homeobox and leucine zipper protein Homez, KIAA1443Homeodomain leucine zipper-containing factor | |
| HOMEZ (AAH65835.1, 1 a.a. - 526 a.a.) full-length human protein. MPPNKEASGLSSSPAGLICLPPISEELQLVWTQAAQTSELDSNEHLLKTFSYFPYPSLADIALLCLRYGLQMEKVKTWFMAQRLRCGISWSSEEIEETRARVVYRRDQLHFKSLLSFTHHAGRPPEEVPPPPVPAPEQVGIGIGPPTLSKPTQTKGLKVEPEEPSQMPPLPQSHQKLKESLMTPGSGAFPYQSDFWQHLQSSGLSKEQAGRGPNQSHGIGTASWNHSTTVPQPQARDKPPPIALIASSCKEESASSVTPSSSSTSSSFQVLANGATAASKPLQPLGCVPQSVSPSEQALPPHLEPAWPQGLRHNSVPGRVGPTEYLSPDMQRQRKTKRKTKEQLAILKSFFLQCQWARREDYQKLEQITGLPRPEIIQWFGDTRYALKHGQLKWFRDNAVPGAPSFQDPAIPTPPPSTRSLNERAETPPLPIPPPPPDIQPLERYWAAHQQLRETDIPQLSQASRLSTQQVLDWFDSRLPQPAEVVVCLDEEEEEEEEELPEDDEEEEEEEEEDDDDDDDDVIIQD | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.4) | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 57594 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction