missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Homez Antibody (PCRP-HOMEZ-1B5), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-08303CL1
This item is not returnable.
View return policy
Description
Homez Monoclonal antibody specifically detects Homez in Human samples. It is validated for Flow Cytometry,Immunohistochemistry (Paraffin),Protein Array
Specifications
| Homez | |
| Monoclonal | |
| CoraFluor 1 | |
| homeobox and leucine zipper encoding, homeobox and leucine zipper protein Homez, KIAA1443Homeodomain leucine zipper-containing factor | |
| Recombinant full-length human Homez protein (Uniprot: Q8IX15) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry (Paraffin), Protein Array | |
| PCRP-HOMEZ-1B5 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 57594 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction