missing translation for 'onlineSavingsMsg'
Learn More

Homogentisic Acid gamma-Lactone, TRC

Product Code. 30404647
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404647 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30404647 Supplier Toronto Research Chemicals Supplier No. TRCH5934101G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-hydroxy-3H-1-benzofuran-2-one
CAS 2688-48-4
Molecular Formula C8 H6 O3
Quantity 1 g
Synonym 5-Hydroxy-2(3H)-benzofuranone, (2,5-Dihydroxyphenyl)-acetic Acid γ-Lactone, 2,5-Dihydroxyphenylacetic Acid Lactone, 2,5-Dihydroxyphenylacetic Acid γ-Lactone, 5-Hydroxy-2(3H)-benzofuranone, 5-Hydroxy-2-coumaranone, 5-Hydroxy-3H-benzofuran-2-one, Homogentisic Acid Lactone, Homogentisic Lactone, NSC 61996
InChI Formula InChI=1S/C8H6O3/c9-6-1-2-7-5(3-6)4-8(10)11-7/h1-3,9H,4H2
SMILES Oc1ccc2OC(=O)Cc2c1
IUPAC Name 5-hydroxy-3H-1-benzofuran-2-one
Molecular Weight (g/mol) 150.131
Formula Weight 150.032
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.