Learn More
Honeywell Analytics™ GasAlertClip Extreme Single-Gas Detectors
Product Code. 15620515
Change view
Click to view available options
Components Measured:
Oxygen
Measurement Range:
0 to 30%
Warranty:
2 Years
Battery Life:
2 Years
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Components Measured Measurement Range Warranty Battery Life Unit Size
15620515 Oxygen 0 to 30% 2 Years 2 Years Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15620515 Supplier Honeywell Analytics™ Supplier No. BWC2X
  • ONLINE EXCLUSIVE PRICE
    € 416.11
    € 249.60 / Pack of 1
    Save € 166.51
    40% Off
Estimated Shipment: 19-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Durable, reliable, accurate, easy to use
Simplicity and minimal training requirements make these gas detectors low-cost option and compact, lightweight design makes them versatile and comfortable.
  • Full function self-test of sensor, battery status, circuit integrity and audible/visual alarms
  • Bright wide-angled visual alarm bars and internal vibrating alarm for high noise environments
  • Large confidence LCD displays life remaining in months, days and hours
  • Water resistant
SO2
  • Temperature Range: -30° to +50°C (-22° to +122°F)
  • Measuring Range: 0 to 100ppm
  • Alarm Level (low/high): 5/10ppm
  • Warranty: Two-year detector only
O2
  • Temperature Range: -20° to +50°C (-4° to +122°F)
  • Measuring Range: 0 to 30.0% by vol
  • Alarm Level (low/high): 19.5/23.5%
  • Requires calibration
  • Warranty: Two-year detector only
CO
  • Temperature Range: -30° to +50°C (-22° to +122°F)
  • Measuring Range: 0 to 300ppm
  • Alarm Level (low/high): 35/200ppm
  • Two- and three-year detectors available
H2S
  • Alarm Level (low/high): Selectable to 5/10ppm or 10/15ppm
  • Temperature Range: -40° to +50°C (-40° to +122°F)
  • Two- and three-year detectors available
  • Measuring Range: 0 to 100ppm
Applications:
For detection of H2S, CO, SO2, O2
Compliance
ATEX: CE III 1 G Exia IIC T4, CE: European Conformity, Class I, Division 1, Group A, B, C, D, IECEx: Exia IIC T4, INMETRO: BR Exia IIC T4, EMC Directive 89/336/EEC
Warranty
Two- or three-year warranty from activation (given normal operation plus one year shelf life)
Specifications
Alarm Types Visual, vibrating, audible (95 dB)
Battery Life 2 Years
Certifications/Compliance ATEX: CE III 1 G Exia IIC T4, CE: European Conformity, Class I, Division 1, Group A, B, C, D, IECEx: Exia IIC T4, INMETRO: BR Exia IIC T4, EMC Directive 89/336/EEC
Depth (English) 1.1 in.
Depth (Metric) 2.79 cm
Components Measured Oxygen
Display Type LCD
Height (English) 3.2 in.
Height (Metric) 8.12 cm
Humidity Range 5 to 95%
IP Rating IP66/67
Length (Metric) 5.08 cm
Measurement Range 0 to 30%
Temperature Range (English) -4°F to +122°F
Temperature Range (Metric) -20°C to +50°C
Warranty 2 Years
Weight (English) 2.7 oz.
Weight (Metric) 76g
Width (English) 2 in.
Width (Metric) 5.08 cm