Learn More
Honeywell™ Txl Pionner S3 Shoes
Brand: Honeywell™ 6244411-46
Additional Details : Weight : 0.01000kg
Specifications
EN ISO 20345:2011, SRC Certified | |
Textile | |
46 | |
Double density Polyurethane | |
Hydrocarbures, sole heat resistant up to 120°C, very good abrasion resistance | |
Shoes |
For professional use such as industry, green spaces, logistics | |
Leather | |
Full, ergonomical, removable and changeable insole, FLEXIUM™ composite puncture resistant mid-sole | |
Freelium Non-metallic | |
Yes |