missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOOK2 Recombinant Rabbit Monoclonal Antibody (JE61-22)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544863
This item is not returnable.
View return policy
Description
HOOK2 Recombinant Monoclonal Antibody for Western Blot, IHC (P), Flow
Hook proteins are cytosolic coiled-coil proteins that contain conserved N-terminal domains, which attach to microtubules, and more divergent C-terminal domains, which mediate binding to organelles. The Drosophila Hook protein is a component of the endocytic compartment.
Specifications
| HOOK2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q7TMK6, Q96ED9 | |
| HOOK2 | |
| Synthetic peptide within human HOOK2 aa 13-62/719. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| JE61-22 | |
| Unconjugated | |
| HOOK2 | |
| A630054I03Rik; hHK2; h-hook2; HK2; hook homolog 2; hook homolog 2 (Drosophila); hook microtubule tethering protein 2; hook microtubule-tethering protein 2; HOOK2; mHK2; protein Hook homolog 2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 170833, 29911, 304669 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction