missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOP Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97809PCP
This item is not returnable.
View return policy
Description
HOP Polyclonal antibody specifically detects HOP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| HOP | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| CAMEO, HOD, homeodomain-only protein, HOP homeobox, HOPLAGYNECC1OB1SMAP31, Lung cancer-associated Y protein, MGC20820, not expressed in choriocarcinoma clone 1, Not expressed in choriocarcinoma protein 1, odd homeobox 1 protein, Odd homeobox protein 1, TOTO | |
| Produced in rabbits immunized with E. coli-derived Human HOP fragment. | |
| 0.1 mL | |
| Cancer | |
| 84525 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction