missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HORMAD2 Antibody (OTI3E4), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72416H
This item is not returnable.
View return policy
Description
HORMAD2 Monoclonal specifically detects HORMAD2 in Human samples. It is validated for Western Blot.
Specifications
| HORMAD2 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| HORMAD2 | |
| Full length recombinant protein of human HORMAD2 (NP_689723) produced in HEK293T cells. | |
| 0.1 mL | |
| 150280.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI3E4 | |
| Western Blot | |
| HORMA domain containing 2, HORMA domain-containing protein 2, MGC26710 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction