missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hormone-sensitive Lipase/HSL Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76735H
This item is not returnable.
View return policy
Description
Hormone-sensitive Lipase/HSL Polyclonal antibody specifically detects Hormone-sensitive Lipase/HSL in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Hormone-sensitive Lipase/HSL | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| hormone-sensitive lipase, hormone-sensitive lipase testicular isoform, HSLEC 3.1.1.79, LHS, lipase, hormone-sensitive | |
| Antibody was raised against a 14 amino acid synthetic peptide from near the carboxy terminus of human Lipe. The immunogen is located within amino acids 920 - 970 of Lipe. Amino Acid Squence: FPEGFHPRRSSQGA | |
| 0.1 mL | |
| Cancer, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Lipid Droplets, Signal Transduction | |
| 3991 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction