Learn More
Welch Ilmvac™ Hose Adapter Kit
Brand: Welch Ilmvac™ 404004
Additional Details : Weight : 0.50000kg
Specifications
Hose Connector, Centering Ring, Hinge Clamp Ring And 2.5m Vacuum Hose, Rubber Red. | |
1 Pack | |
DN 25 KF - DN 8 | |
Hose Adapter Kit |