Learn More
Buerkle™ Hose Angle Bracket with Strong Outer Thread
Product Code. 10657331 Shop All Bürkle Products
Click to view available options
Diameter (Metric):
12 mm nozzle
25 mm nozzle
Nozzle: 20 mm
Nozzle: 22 mm
Thread Size:
1 in.
1/2 in.
3/4 in.
Unit Size:
Each
Product Code. 10657331
Brand: Buerkle™ 85470100
Only null left
Add to basket
Add to basket
Angled at 90°.
  • Manufactured from polypropylene
  • Fixed
  • External thread
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EU Foodstuffs
Diameter (Metric) 25 mm nozzle
Autoclavable Autoclavable
Material Polypropylene
For Use With Ball Valve
Thread Size 1 in.
Width (Metric) 19.5 mm
Product Type Tube Bend