missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Host cell factor C2 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99120R
This item is not returnable.
View return policy
Description
Host cell factor C2 Polyclonal antibody specifically detects Host cell factor C2 in Human samples. It is validated for Western Blot
Specifications
| Host cell factor C2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 29915 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 550 | |
| FLJ94012, HCF2, HCF-2C2 factor, host cell factor 2, host cell factor C2 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Host cell factor C2. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction