Learn More
Honeywell™ Howard Leight™ Viking™ Ear Muffs
Catalog Number p-3202011
Click to view available options
Noise Reduction Rating (NRR):
29 dB
Style:
Viking 1 Earmuffs
Wear over/behind-the-head or under-the-chin
  • Fits with other PPE, including hard hats, visors, face shields, respirators and welding shields
  • Dielectric construction won't deform
  • Snap-in ear cushions
TRUSTED_SUSTAINABILITY
Includes Inner ventilation
Certifications/Compliance ANSI S.3.19-1974
For Use With (Application) Agriculture and farming, building construction, forestry, general contracting, heavy construction, landscaping, lumber or wood products, metal fabrication
Petrochemical
Steel
Mining
Transportation
Utility/Waste Management
Noise Reduction Rating (NRR) 29 dB
Product Type Ear Muffs
Dielectric Yes
Style Viking 1 Earmuffs