missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXA11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA579387
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Brain Tissue, Human Placenta Tissue, HLEA whole cell, HT1080 whole cell, HEPA whole cell. Flow: U937 cell.
HOX11, also called homeobox11 or Tcl-3 is located at the 10q24 T-Cell translocation breakpoint region. It encodes a homeobox-domain containing protein, HOX11, containing a glycine and proline-rich amino terminus. HOX11 is required for maintenance of the developing spleen and cell survival. The HOX11protein binds to a specific DNA sequence and localizes to the cell nucleus. It transactivates transcription of a reporter gene linked to a cis-regulatory element, acting as a positive transcription activator. The HOX11 gene is highly expressed in T-ALLs as a result of a t (7:10) (q35;q24) or t (10:14) (q24;qll) translocation. The homeobox gene is deregulated upon translocation into the T-cell receptor locus and activates a specific subset of tumor-associated target genes. HOX 11 gene translocation has been attributed to T-cell leukemia and lymphoid neoplasias.
Spezifikation
| HOXA11 | |
| Polyclonal | |
| Unconjugated | |
| HOXA11 | |
| homeo box 1I; homeo box A11; homeobox A11; homeobox protein Hox-1.9; Homeobox protein Hox-1I; homeobox protein HOXA11; homeobox protein Hox-A11; HOX1; Hox-1.9; HOX11; HOX1I; Hoxa11; Hoxa-11; NEWGENE_1564605; RUSAT1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 103692131, 15396, 3207 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| P31270, P31311 | |
| HOXA11 | |
| E.coli-derived human HOXA11 recombinant protein (Position: H74-R245). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur