missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXA3 Monoclonal Antibody (OTI8C5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811549S
This item is not returnable.
View return policy
Description
HOXA3 Monoclonal antibody specifically detects HOXA3 in Human samples. It is validated for Western BlotSpecifications
| HOXA3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P23276 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3792 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI8C5 | |
| Unconjugated | |
| KEL | |
| homeo box 1E; homeo box A3; homeobox A2; homeobox A3; homeobox protein Hox-1.5; Homeobox protein Hox-1E; homeobox protein Hox-A3; Homeobox protein MO-10; HOX1; Hox-1.5; Hox-1.5-like protein; Hox-15; HOX1E; HOXA3; Hoxa-3; Mo-10; NEWGENE_1561431 | |
| Human recombinant protein fragment corresponding to amino acids 166-420 of human KEL produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction