missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HOXA7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15597581
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15597581 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15597581 Supplier Invitrogen™ Supplier No. PA536245

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 25 kDa. Purity is >95% by SDS-PAGE.

Hox genes play a fundamental role in the development of the vertebrate central nervous system, heart, axial skeleton, limbs, gut, urogenital tract and external genitalia. HoxA2 protein may contribute to dorsal-ventral patterning and/or to the specification of neuronal phenotype. HoxA7 functions as a potent transcriptional repressor and its action as such requires several domains, including both activator and repressor regions. HoxA7 is expressed in the fetal liver, lung, skeletal muscle, kidney, pancreas and placenta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXA7
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide; pH 7.2
Gene HOXA7
Gene Accession No. P02830, P09634, P31268
Gene Alias ANTP; AV118143; homeo box A7; homeobox A7; homeobox A9; Homeobox gene A7; homeobox protein Hox 1.1; Homeobox protein Hox-1.1; homeobox protein HOX-1A; Homeobox protein Hox-A7; HOMEOBOX PROTEIN HOX-A7 (HOX-1.1) (M6-12) (M6); Homeobox protein M6-12; homeobox protein R5; HOX1; HOX1.1; hox-1.1; HOX1A; Hox1r5; Hoxa7; Hoxa-7; Hoxa9; M6
Gene Symbols HOXA7
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 100-150 of human Hox-A7.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15404, 3204, 500126
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.