missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXA9 Recombinant Rabbit Monoclonal Antibody (23GB6645), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553087
This item is not returnable.
View return policy
Description
HOXA9 is the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development.
Specifications
| HOXA9 | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P31269 | |
| Hoxa9 | |
| A synthesized peptide derived from human HOXA9 (231-272AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB6645 | |
| Unconjugated | |
| Hoxa9 | |
| ABD-B; D6a9; homeo box A9; homeobox A9; homeobox A9-like; homeobox protein Hox-1.7; Homeobox protein Hox-1G; homeobox protein Hox-A9; homeodomain protein HOXA9; HOX1; HOX1.7; Hox-1.7; HOX1G; HOXA 9; Hoxa9; Hoxa-9; Hoxa-9 protein; Hoxa9l; RGD1310001 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3205 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction