missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Hoxb1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568959
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 79%; Dog: 86%; Horse: 86%; Human: 100%; Mouse: 79%; Pig: 93%; Rat: 86%.
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis. Acts on the anterior body structures.
Specifications
| Hoxb1 | |
| Polyclonal | |
| Unconjugated | |
| Hoxb1 | |
| HCFP3; homeo box B1; homeobox B1; homeobox protein Hox-2.9; Homeobox protein Hox-2I; homeobox protein Hox-B1; HOX2; Hox-2.9; Hox-29; HOX2I; Hoxb1; Hoxb-1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 303491 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| Hoxb1 | |
| synthetic peptide directed towards the N-terminal region of rat Hoxb1. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction