missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXB7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12613-1-AP-20UL
This item is not returnable.
View return policy
Description
The homeobox genes encode transcription factors that regulate developmental processes such as body patterning and organogenesis. The mammalian HOX gene complex consists of 39 genes that are located on 4 linkage groups (clusters), dispersed over 4 chromosomes (HOXA, HOXB, HOXC and HOXD). HOXB7 is expressed in most maligt T-cell lines and CD4-positive peripheral blood cells, and is constitutively expressed in both melanoma primary lesions and cell lines. HOXB7 binds to and transactivates bFGF in melanoma cell lines but SkBr3 breast adenocarcinoma is negative for the expression of both HOXB7 and bFGF genes. HOXB7 may be a key activator of tumor-associated neoangiogenesis.Specifications
| HOXB7 | |
| Polyclonal | |
| Unconjugated | |
| HOXB7 | |
| HHO.C1, homeobox B7, Hox 2.3, HOX2, HOX2C, HOXB7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15415, 3217, 497985 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P09024, P09629, P18864 | |
| HOXB7 | |
| HOXB7 Fusion Protein Ag3245 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction