missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXB8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98722-50ul
This item is not returnable.
View return policy
Description
HOXB8 Polyclonal antibody specifically detects HOXB8 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| HOXB8 | |
| Polyclonal | |
| Western Blot 1:500-1:1000, Immunoprecipitation 1-2 μL/mg of lysate | |
| homeo box 2D, homeo box B8, homeobox B8, Homeobox protein Hox-2.4, Homeobox protein Hox-2D, homeobox protein Hox-B8, HOX2, HOX2DHox-2.4 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human HOXB8. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 3218 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction