missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXB8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103891
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HOXB8.
This gene is a member of the Antp homeobox family and encodes a nuclear protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox B genes located on chromosome 17. The encoded protein functions as a sequence-specific transcription factor that is involved in development. Increased expression of this gene is associated with colorectal cancer. Mice that have had the murine ortholog of this gene knocked out exhibit an excessive pathologic grooming behavior. This behavior is similar to the behavior of humans suffering from the obsessive-compulsive spectrum disorder trichotillomania.
Specifications
| HOXB8 | |
| Polyclonal | |
| Unconjugated | |
| HOXB8 | |
| homeo box 2D; homeo box B8; homeobox B8; homeobox gene B8; homeobox protein Hox-2.4; Homeobox protein Hox-2D; homeobox protein Hox-B8; homeobox protein R1a; HOX2; Hox-2.4; Hox2.4 homeobox homolog; HOX2D; Hox2r1a; Hoxb8; Hoxb-8 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15416, 24457, 3218 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P09632, P17481, P18863 | |
| HOXB8 | |
| A synthesized peptide derived from human HOXB8(Accession P17481), corresponding to amino acid residues P61-K111. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction