missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXC10 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71933DL594
This item is not returnable.
View return policy
Description
HOXC10 Polyclonal antibody specifically detects HOXC10 in Human samples. It is validated for Western Blot
Specifications
| HOXC10 | |
| Polyclonal | |
| Western Blot | |
| homeo box C10, homeobox C10, Homeobox protein Hox-3I, homeobox protein Hox-C10, homeoprotein C10, HOX3I, MGC5259 | |
| The immunogen for this product maps to a region between residue 100 and 150 of human Homeobox C10 using the numbering given in entry NP_059105.2 (GeneID 3226). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 3226 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction