Learn More
HOXC10 Monoclonal Antibody (OTI1E10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA808890
Encompass_Preferred
 View more versions of this product
Product Code. 17580634
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The protein level is controlled during cell differentiation and proliferation, which may indicate this protein has a role in origin activation.
Specifications
Specifications
HOXC10
Monoclonal
Unconjugated
HOXC10
HOX3I
Mouse
Affinity Chromatography
RUO
3226
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
OTI1E10
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9NYD6
HOXC10
Human recombit protein fragment corresponding to amino acids 1-262 of human HOXC10 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2a