missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXC10 Monoclonal Antibody (OTI1H5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808866S
This item is not returnable.
View return policy
Description
HOXC10 Monoclonal antibody specifically detects HOXC10 in Human samples. It is validated for Western BlotSpecifications
| HOXC10 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NYD6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3226 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1H5 | |
| Unconjugated | |
| HOXC10 | |
| AI506621; homeo box C10; homeobox C10; homeobox protein Hox-3.6; Homeobox protein Hox-3I; homeobox protein Hox-C10; homeoprotein C10; Hox3.5 homeobox; Hox-3.6; HOX3I; HOXC10; Hoxc-10 | |
| Human recombinant protein fragment corresponding to amino acids 1-262 of human HOXC10 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction