Learn More
HOXC10 Monoclonal Antibody (OTI5D4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA808881S
Encompass_Preferred
 View more versions of this product
Product Code. 17560614
  • € 242.00 / 30µL
Description
HOXC10 Monoclonal antibody specifically detects HOXC10 in Human samples. It is validated for Western Blot
Specifications
Specifications
HOXC10
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9NYD6
Mouse
Affinity chromatography
RUO
3226
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI5D4
Unconjugated
HOXC10
AI506621; homeo box C10; homeobox C10; homeobox protein Hox-3.6; Homeobox protein Hox-3I; homeobox protein Hox-C10; homeoprotein C10; Hox3.5 homeobox; Hox-3.6; HOX3I; HOXC10; Hoxc-10
Human recombinant protein fragment corresponding to amino acids 1-262 of human HOXC10 produced in E.coli.