missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXC11 Antibody (OTI3E10), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70913B
This item is not returnable.
View return policy
Description
HOXC11 Monoclonal specifically detects HOXC11 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| HOXC11 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| HOXC11 | |
| Recombinant protein fragment corresponding to amino acids 1-304 of human HOXC11 (NP_055027) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI3E10 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| homeo box C11, homeobox C11, Homeobox protein Hox-3H, homeobox protein Hox-C11, HOX3H, MGC4906 | |
| Mouse | |
| Affinity Purified | |
| DNA replication Transcription Translation and Splicing | |
| 3227.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction