missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXC11 Antibody (OTI3E10), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70913MFV500
This item is not returnable.
View return policy
Description
HOXC11 Monoclonal antibody specifically detects HOXC11 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| HOXC11 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| OTI3E10 | |
| Flow Cytometry | |
| homeo box C11, homeobox C11, Homeobox protein Hox-3H, homeobox protein Hox-C11, HOX3H, MGC4906 | |
| Human recombinant protein fragment corresponding to amino acids 1-304 of human HOXC11(NP_055027) produced in E.coli. | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 3227 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction