missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXC4 Monoclonal Antibody (OTI2A5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809725S
This item is not returnable.
View return policy
Description
HOXC4 Monoclonal antibody specifically detects HOXC4 in Human samples. It is validated for Western BlotSpecifications
| HOXC4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BY41 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55869 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI2A5 | |
| Unconjugated | |
| HDAC8 | |
| cp19; homeo box 3E; homeo box C4; homeobox C4; Homeobox gene C4; homeobox protein CP19; Homeobox protein Hox-3.5; homeobox protein Hox-3E; Homeobox protein Hox-C4; homeobox protein R3; HOX3; Hox-3.5; Hox-35; HOX3E; Hox3r3; Hoxc4; Hoxc-4 | |
| Human recombinant protein fragment corresponding to amino acids 1-243 of human HDAC8 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction