missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXD10 Polyclonal Antibody

Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene is a member of the Abd-B homeobox family and encodes a protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox D genes located on chromosome 2. The encoded nuclear protein functions as a sequence-specific transcription factor that is expressed in the developing limb buds and is involved in differentiation and limb development. Mutations in this gene have been associated with Wilm's tumor and congenital vertical talus and/or a foot deformity resembling that seen in Charcot-Marie-Tooth disease.
Specifications
Specifications
| Antigen | HOXD10 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with trehalose and no preservative |
| Gene | HOXD10 |
| Gene Accession No. | P28358 |
| Gene Alias | AI385591; AI874987; d10; homeo box 4D; homeo box D10; homeo box D10a; homeobox D10; homeobox D10a; homeobox gene D-10; homeobox protein Hox-4.5; Homeobox protein Hox-4D; homeobox protein Hox-4E; Homeobox protein Hox-5.3; homeobox protein Hox-C10; Homeobox protein Hox-D10; homeobox protein Hox-D10a; homeobox protein Hox-D11; homeobox protein Hox-D10; HOX4; Hox-4.4; Hox-4.5; Hox-45; HOX4D; HOX4E; Hox-5.3; hoxc10; hox-C10; Hoxc10 protein; Hoxd10; HOX-D10; hoxd-10; hoxd10a; hoxd10a protein; z-82 |
| Gene Symbols | HOXD10 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?