missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXD8 Mouse anti-Human, Clone: OTI2A1, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810942S
This item is not returnable.
View return policy
Description
HOXD8 Monoclonal antibody specifically detects HOXD8 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| HOXD8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P13378 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3234 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A1 | |
| Unconjugated | |
| HOXD8 | |
| 4921540P06Rik; AI047735; homeo box 4E; homeo box D8; homeobox D8; homeobox protein 5.4; homeobox protein Hox-4.3; Homeobox protein Hox-4E; homeobox protein Hox-5.4; Homeobox protein Hox-D8; homeobox protein Hox-D8-like; HOX4; Hox-4.3; Hox-4.5; HOX4E; HOX5.4; Hox-5.4; HOXD8; Hoxd-8; LOC100912936 | |
| Human recombinant protein fragment corresponding to amino acids 69-290 of human HOXD8 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction