Learn More
HOXD9 Monoclonal Antibody (OTI2B1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809665
Encompass_Preferred
 View more versions of this product
Product Code. 17542114
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
HOXD9 is the sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located at 2q31-2q37 chromosome regions. Deletions that removed the entire HOXD gene cluster or 5′ end of this cluster have been associated with severe limb and genital abnormalities. The exact role of this gene has not been determined.
Specifications
Specifications
HOXD9
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P28356
HOXD9
Human recombit protein fragment corresponding to amino acids 109-352 of human HOXD9 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI2B1
Unconjugated
HOXD9
HOX4, HOX4C, Hox-4.3, Hox-5.2
Mouse
Affinity Chromatography
RUO
3235
-20° C, Avoid Freeze/Thaw Cycles