missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXD9 Monoclonal Antibody (OTI4H10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF809664
This item is not returnable.
View return policy
Description
HOXD9 Monoclonal antibody specifically detects HOXD9 in Human samples. It is validated for Western BlotSpecifications
| HOXD9 | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P15408 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 2355 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4H10 | |
| Unconjugated | |
| FOSL2 | |
| homeo box D9; homeobox D9; homeobox Hox-D9; homeobox protein Hox-4.4; Homeobox protein Hox-4C; homeobox protein Hox-5.2; Homeobox protein Hox-D9; HOX4; Hox-4.3; Hox-4.3, mouse, homolog of; Hox-4.4; HOX4C; Hox-5.2; HOXD9; Hoxd-9 | |
| Human recombinant protein fragment corresponding to amino acids 110-326 of human FOSL2 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction