missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HP1 alpha Antibody (RAB-C133) - F(ab) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
€ 399.00 - € 849.00
Specifications
| Antigen | HP1 alpha |
|---|---|
| Clone | RAB-C133 |
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunoprecipitation, Mass Spectrometry, ChIP Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18659953
|
Novus Biologicals
NBP3-20126-0.025mg |
0.025 mg |
€ 399.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18644834
|
Novus Biologicals
NBP3-20126-0.1mg |
0.1 mg |
€ 849.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HP1 alpha Monoclonal antibody specifically detects HP1 alpha in Human samples. It is validated for ELISA, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)Specifications
| HP1 alpha | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Antigen p25, chromobox homolog 5, chromobox homolog 5 (Drosophila HP1 alpha), chromobox homolog 5 (HP1 alpha homolog, Drosophila), chromobox protein homolog 5, Heterochromatin protein 1 homolog alpha, heterochromatin protein 1-alpha, HP1, HP1 alpha, HP1 alpha homolog, HP1A, HP1-ALPHA, HP1Hs alpha, HP1Hs-alpha | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing CBX5 protein under non-denaturing conditions. | |
| Affinity purified |
| RAB-C133 | |
| ELISA, Immunoprecipitation, Mass Spectrometry, ChIP Assay | |
| Unconjugated | |
| Human | |
| Cellular Markers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing | |
| PBS | |
| 23468 | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title