Learn More
HP1 alpha Antibody (RAB-C133), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-20127PCP
Product Code. 18613895
  • € 462.00 / 0.10mL
Description
HP1 alpha Monoclonal antibody specifically detects HP1 alpha in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunoprecipitation
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, Chromatin Immunoprecipitation (ChIP)
Rabbit
PerCP
Antigen p25, chromobox homolog 5, chromobox homolog 5 (Drosophila HP1 alpha), chromobox homolog 5 (HP1 alpha homolog, Drosophila), chromobox protein homolog 5, Heterochromatin protein 1 homolog alpha, heterochromatin protein 1-alpha, HP1, HP1 alpha, HP1 alpha homolog, HP1A, HP1-ALPHA, HP1Hs alpha, HP1Hs-alpha
23468