Learn More
HP1 alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NB110-40623
This item is not returnable.
View return policy
Description
HP1 alpha Polyclonal specifically detects HP1 alpha in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Polyclonal | |
| IgG |