missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ HP1 alpha Protein

Product Code. 18492701 Shop All Bio Techne Products
Click to view available options
Quantity:
0.5 mg
100μg
Unit Size:
0.50mg
100µg
This item is not returnable. View return policy

Product Code. 18492701

Brand: Novus Biologicals™ NBP1988630.5mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-191 of Human HP1 alpha The Recombinant Human HP1 alpha Protein is derived from E. coli. The Recombinant Human HP1 alpha Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_001120794
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. In 20 mM Tris-HCl buffer (pH 8.0) containing 1 mM DTT, 30% glycerol, 0.1 M NaCl
Gene ID (Entrez) 23468
Molecular Weight (g/mol) 22 kDa
Purification Method SDS-PAGE
Quantity 0.5 mg
Source Recombinant human CBX5 protein, fused to His-tag at N-terminus, expressed in E. coli
Immunogen CB x 5, 1-191 aa, Human, His tag, E. coli (NP_001120794).
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name HP1 alpha
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.