missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HP1BP3 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18487JF646
This item is not returnable.
View return policy
Description
HP1BP3 Polyclonal antibody specifically detects HP1BP3 in Human samples. It is validated for Western Blot
Specifications
| HP1BP3 | |
| Polyclonal | |
| Western Blot | |
| heterochromatin protein 1, binding protein 3, heterochromatin protein 1-binding protein 3, HP1-BP74, MGC43701, Protein HP1-BP74 | |
| Recognizes region between residues Between between 503 and 553.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 50809 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction