missing translation for 'onlineSavingsMsg'
Learn More

IDEXX Water HPC for Quanti-Tray

IDEXX Water HPC for Quanti-Tray is an accurate test for determining heterotrophic plate counts in a 100ml sample.

Brand:  IDEXX Water 98-27389-00

Additional Details : Weight : 0.08600kg

 View more versions of this product

Product Code. 15941190

  • POA / 20 tests

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

  • Heterotrophic plate count made easy for bottling and manufacturing plants and on-premise testing
  • Quantifies heterotrophic organisms in a 100 mL format
  • No autoclaving, filtering, or agar
  • No colony counting or interpretation
  • Convenient 100-mL sample size—test source water, transport vehicle water, manufacturing line water, and finished products
  • Simple procedure needs no special training
  • Shares the same Quanti-Tray system as Colilert, Enterolert, and Pseudalert tests for maximum efficiency
  • Results valid at 44–72 hours
Specifications

Specifications

Pink
20/Pk.
Store at 2 to 30°C, away from light
44 hr.
Quanti-Tray
100 mL
Quantification
Heterotrophic plate count
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers