Learn More
HPCAL4 Monoclonal Antibody (OTI2A12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA808693
Encompass_Preferred
 View more versions of this product
Product Code. 17500204
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
May be involved in the calcium-dependent regulation of rhodopsin phosphorylation.
Specifications
Specifications
HPCAL4
Monoclonal
Unconjugated
HPCAL4
HLP4
Mouse
Affinity Chromatography
RUO
51440
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
OTI2A12
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9UM19
HPCAL4
Human recombit protein fragment corresponding to amino acids 2-191 of human HPCAL4 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2a