missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HPCAL4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565824
This item is not returnable.
View return policy
Description
Immunogen sequence: EMLEIIEAIYK MVGTVIMMRM NQDGL Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
May be involved in the calcium-dependent regulation of rhodopsin phosphorylation.
Specifications
| HPCAL4 | |
| Polyclonal | |
| Unconjugated | |
| HPCAL4 | |
| AI846570; hippocalcin like 4; hippocalcin-like 4; hippocalcin-like protein 4; HLP4; HPCAL4; Neural visinin-like protein 2; NVP-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51440 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UM19 | |
| HPCAL4 | |
| Recombinant Human HPCAL4. Recombinant protein control fragment (Product #RP-105627). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction