Learn More
HPGD Mouse anti-Human, Clone: OTI2C10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF502561
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Prostaglandin inactivation. Contributes to the regulation of events that are under the control of prostaglandin levels. Catalyzes the NAD-dependent dehydrogenation of lipoxin A4 to form 15-oxo-lipoxin A4. Inhibits in vivo proliferation of colon cancer cells.Specifications
| HPGD | |
| Monoclonal | |
| Unconjugated | |
| HPGD | |
| 15-PGDH, PGDH, PGDH1, PHOAR1, SDR36C1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3248 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2C10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P15428 | |
| HPGD | |
| Full length human recombit protein of human HPGD produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.