Learn More
hPGDS-IN-1, MedChemExpress
Product Code. 16478067
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478067 5 mg 5mg
16421547 50 mg 50mg
16405358 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16478067 Supplier Medchem Express Supplier No. HY12791
MedChemExpress hPGDS-IN-1 is a hPGDS inhibitor ,with IC50 of 12 nM in the Fluorescence Polarization Assay or the EIA assay.
hPGDS-IN-1 is a hPGDS inhibitor ,with IC50 of 12 nM in the Fluorescence Polarization Assay or the EIA assay.
Specifications
Chemical Name or Material hPGDS-IN-1
CAS 1234708-04-3
Color White
Purity Grade Notes Research
Molecular Formula C22H20N6O3
Quantity 5 mg
Solubility Information DMSO : ≥ 30 mg/mL (72.04 mM)
SMILES OC(C)(C)C1=NC(C2=CC(CNC(C(C=N3)=CN=C3C4=CC=CC=N4)=O)=CC=C2)=NO1
Molecular Weight (g/mol) 416.43
Formula Weight 416.43
Percent Purity 99.13%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only