missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPi1 Antibody (HIC0-4F9), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-18872JF549
This item is not returnable.
View return policy
Description
HPi1 Monoclonal antibody specifically detects HPi1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
| HPi1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Mouse | |
| Tissue culture supernatant | |
| RUO | |
| Human, Mouse | |
| Supernatant |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Frozen) | |
| HIC0-4F9 | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Frozen | |
| human pancreatic endocrine islet cell | |
| Human pancreatic enriched islet cells containing low levels of exocrine and ductal cells. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction